Alfa Aesar™ N-Benzyloxycarbonyl-L-phenylalaninol, 98%

Manufacturer: Alfa Aesar™ B2503614

Quantity 25g

 View more versions of this product

Catalog No. AAB2503614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6372-14-1
    Molecular Formula C17H19NO3
    Formula Weight 285.35
    MDL Number MFCD00191138
    Synonym (S)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; N-Cbz-L-phenylalaninol
    Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninol

    Specifications

    Formula Weight 285.35
    CAS 6372-14-1
    Molecular Formula C17H19NO3
    Melting Point 92° to 95°C
    MDL Number MFCD00191138
    Quantity 25g
    Synonym (S)-2-Carbobenzyloxyamino-3-phenyl-1-propanol; N-Cbz-L-phenylalaninol
    Chemical Name or Material N-Benzyloxycarbonyl-L-phenylalaninol
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit