Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
40291-26-7
C17H16N2O7
360.322
MFCD00191144
LZNYDJDSWANAND-LBPRGKRZSA-N
z-pyr-osu,s-1-benzyl 2-2,5-dioxopyrrolidin-1-yl 5-oxopyrrolidine-1,2-dicarboxylate,1-benzyl 2-2,5-dioxopyrrolidin-1-yl 2s-5-oxopyrrolidine-1,2-dicarboxylate,1-cbz-l-pyroglutamic acid n-succinimidyl ester,z-l-pyroglutamic acid n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl 1-benzyloxy carbonyl-5-oxo-l-prolinate,1-o-benzyl 2-o-2,5-dioxopyrrolidin-1-yl 2s-5-oxopyrrolidine-1,2-dicarboxylate,2s-2-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-5-oxo-1-pyrrolidinecarboxylic acid benzyl ester
1915413
1-O-benzyl 2-O-(2,5-dioxopyrrolidin-1-yl) (2S)-5-oxopyrrolidine-1,2-dicarboxylate
C1CC(=O)N(C1C(=O)ON2C(=O)CCC2=O)C(=O)OCC3=CC=CC=C3
40291-26-7
C17H16N2O7
360.322
MFCD00191144
LZNYDJDSWANAND-LBPRGKRZSA-N
z-pyr-osu,s-1-benzyl 2-2,5-dioxopyrrolidin-1-yl 5-oxopyrrolidine-1,2-dicarboxylate,1-benzyl 2-2,5-dioxopyrrolidin-1-yl 2s-5-oxopyrrolidine-1,2-dicarboxylate,1-cbz-l-pyroglutamic acid n-succinimidyl ester,z-l-pyroglutamic acid n-hydroxysuccinimide ester,2,5-dioxopyrrolidin-1-yl 1-benzyloxy carbonyl-5-oxo-l-prolinate,1-o-benzyl 2-o-2,5-dioxopyrrolidin-1-yl 2s-5-oxopyrrolidine-1,2-dicarboxylate,2s-2-2,5-dioxo-1-pyrrolidinyl oxy carbonyl-5-oxo-1-pyrrolidinecarboxylic acid benzyl ester
1915413
1-O-benzyl 2-O-(2,5-dioxopyrrolidin-1-yl) (2S)-5-oxopyrrolidine-1,2-dicarboxylate
C1CC(=O)N(C1C(=O)ON2C(=O)CCC2=O)C(=O)OCC3=CC=CC=C3
Beilstein 5157479
Formula Weight 360.32
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-L-pyroglutamic acid N-succinimidyl ester

storageNote1 : -30° to -10°C