Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-serine benzyl ester, 99%

Click to view available options
Quantity:
1g
5g
21209-51-8
C18H19NO5
329.352
MFCD00037824
MHHDPGHZHFJLBZ-INIZCTEOSA-N
z-ser-obzl,cbz-ser-obzl,n-cbz-l-serine benzyl ester,n-z-l-serine benzyl ester,s-benzyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate,n-carbobenzoxy-l-serine benzyl ester,benzyl n-benzyloxy carbonyl-l-serinate,l-serine,n-phenylmethoxy carbonyl-, phenylmethyl ester,serine, n-phenylmethoxy carbonyl-, phenylmethyl ester,z-l-serine benzyl ester
6992970
benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
C1=CC=C(C=C1)COC(=O)C(CO)NC(=O)OCC2=CC=CC=C2
21209-51-8
C18H19NO5
329.352
MFCD00037824
MHHDPGHZHFJLBZ-INIZCTEOSA-N
z-ser-obzl,cbz-ser-obzl,n-cbz-l-serine benzyl ester,n-z-l-serine benzyl ester,s-benzyl 2-benzyloxy carbonyl amino-3-hydroxypropanoate,n-carbobenzoxy-l-serine benzyl ester,benzyl n-benzyloxy carbonyl-l-serinate,l-serine,n-phenylmethoxy carbonyl-, phenylmethyl ester,serine, n-phenylmethoxy carbonyl-, phenylmethyl ester,z-l-serine benzyl ester
6992970
benzyl (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoate
C1=CC=C(C=C1)COC(=O)C(CO)NC(=O)OCC2=CC=CC=C2
Applications
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
Melting Point 82°C to 84°C
Quantity 1g
Formula Weight 329.32
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine benzyl ester