Learn More
N-Benzyloxycarbonyl-L-serine benzyl ester, 99%, Thermo Scientific Chemicals
Description
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Benzyloxycarbonyl-L-serine benzyl ester is used as a building block in peptide synthesis and as a biochemical reagent. It is also used to synthesize the corresponding N-benzyloxycarbonyl-O-tert-butyl-(lor dl)-serine benzyl ester. Further, it serves as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 82°C to 84°C |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 329.32 |
Percent Purity | 99% |
Chemical Name or Material | N-Benzyloxycarbonyl-L-serine benzyl ester |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.