Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-serylglycine ethyl ester, 95%

Click to view available options
Quantity:
250mg
1g
5g
4526-93-6
C15H20N2O6
324.333
MFCD00191150
ZOONJGSLKPDXCV-LBPRGKRZSA-N
z-ser-gly-oet,s-ethyl 2-2-benzyloxy carbonyl amino-3-hydroxypropanamido acetate,ethyl 2-2s-2-benzyloxy carbonyl amino-3-hydroxypropanamido acetate,carbobenzoxy-l-seryl glycine ethyl ester,n-benzyloxycarbonyl serylglycine ethyl ester,ethyl 2-2s-3-hydroxy-2-phenylmethoxycarbonylamino propanoyl amino acetate
1742149
ethyl 2-[[(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetate
CCOC(=O)CNC(=O)C(CO)NC(=O)OCC1=CC=CC=C1
4526-93-6
C15H20N2O6
324.333
MFCD00191150
ZOONJGSLKPDXCV-LBPRGKRZSA-N
z-ser-gly-oet,s-ethyl 2-2-benzyloxy carbonyl amino-3-hydroxypropanamido acetate,ethyl 2-2s-2-benzyloxy carbonyl amino-3-hydroxypropanamido acetate,carbobenzoxy-l-seryl glycine ethyl ester,n-benzyloxycarbonyl serylglycine ethyl ester,ethyl 2-2s-3-hydroxy-2-phenylmethoxycarbonylamino propanoyl amino acetate
1742149
ethyl 2-[[(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoyl]amino]acetate
CCOC(=O)CNC(=O)C(CO)NC(=O)OCC1=CC=CC=C1
Beilstein 1893538
Formula Weight 324.33
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-L-serylglycine ethyl ester

storageNote1 : 2° to 8°C