Alfa Aesar™ N-Benzyloxycarbonyl-L-tert-leucine, 99%

Manufacturer: Alfa Aesar™ H27132MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27132MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62965-10-0
    Molecular Formula C14H19NO4
    Formula Weight 265.3
    MDL Number MFCD00137411
    Synonym Z-tert-Leu-OH
    Chemical Name or Material N-Benzyloxycarbonyl-L-tert-leucine

    Specifications

    Formula Weight 265.3
    Molecular Formula C14H19NO4
    MDL Number MFCD00137411
    CAS 62965-10-0
    Quantity 250mg
    Synonym Z-tert-Leu-OH
    Optical Rotation −4° (c=1 in Methanol)
    Chemical Name or Material N-Benzyloxycarbonyl-L-tert-leucine
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit