Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-threoninamide, 97%

Click to view available options
Quantity:
250mg
1g
5g
49705-98-8
C12H16N2O4
252.27
MFCD00038587
PYZXYZOBPGPOFQ-SCZZXKLOSA-N
z-thr-nh2,cbz-l-threoninamide,cbz-thr-nh2,carbobenzoxy-l-threonine amide,n-benzyloxycarbonyl l-threonine amide,z-l-threoninamide,z-l-threonine amide,z-thr-nh,pubchem13626,z-l-thr-nh2
7010574
benzyl N-[(2S,3R)-1-amino-3-hydroxy-1-oxobutan-2-yl]carbamate
CC(C(C(=O)N)NC(=O)OCC1=CC=CC=C1)O
49705-98-8
C12H16N2O4
252.27
MFCD00038587
PYZXYZOBPGPOFQ-SCZZXKLOSA-N
z-thr-nh2,cbz-l-threoninamide,cbz-thr-nh2,carbobenzoxy-l-threonine amide,n-benzyloxycarbonyl l-threonine amide,z-l-threoninamide,z-l-threonine amide,z-thr-nh,pubchem13626,z-l-thr-nh2
7010574
benzyl N-[(2S,3R)-1-amino-3-hydroxy-1-oxobutan-2-yl]carbamate
CC(C(C(=O)N)NC(=O)OCC1=CC=CC=C1)O
Quantity 5g
Color White
Beilstein 2335408
Formula Weight 252.27
Physical Form Powder
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-L-threoninamide

storageNote1 : -30° to -10°C