Alfa Aesar™ N-Benzyloxycarbonyl-L-threonine benzyl ester, 99%

Manufacturer: Alfa Aesar™ B2267603

Quantity 1g

 View more versions of this product

Catalog No. AAB2267603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16597-50-5
    Molecular Formula C19H21NO5
    Formula Weight 343.38
    MDL Number MFCD00077041
    Synonym N-Cbz-L-Threonine benzyl ester; Z-Thr-OBzl
    Chemical Name or Material N-Benzyloxycarbonyl-L-threonine benzyl ester

    Specifications

    Formula Weight 343.38
    Melting Point 78° to 82°C
    Molecular Formula C19H21NO5
    MDL Number MFCD00077041
    CAS 16597-50-5
    Quantity 1g
    Synonym N-Cbz-L-Threonine benzyl ester; Z-Thr-OBzl
    Chemical Name or Material N-Benzyloxycarbonyl-L-threonine benzyl ester
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit