Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-threonine methyl ester, 97%

Click to view available options
Quantity:
5g
25g
100g
57224-63-2
C13H17NO5
267.281
MFCD00038585
OPZWAOJFQFYYIX-KOLCDFICSA-N
z-thr-ome,2s,3r-methyl 2-benzyloxy carbonyl amino-3-hydroxybutanoate,n-carbobenzyloxy-l-threonine methyl ester,z-l-threonine methyl ester,n-carbobenzoxy-l-threonine methyl ester,n-alpha-carbobenzoxy-l-threonine methyl ester,methyl 2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanoate,z-thr-ome,n-benzyloxycarbonyl threonine methyl ester,methyl 2-benzyloxy carbonyl amino-3-hydroxybutanoate #
2733409
methyl (2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoate
CC(C(C(=O)OC)NC(=O)OCC1=CC=CC=C1)O
57224-63-2
C13H17NO5
267.281
MFCD00038585
OPZWAOJFQFYYIX-KOLCDFICSA-N
z-thr-ome,2s,3r-methyl 2-benzyloxy carbonyl amino-3-hydroxybutanoate,n-carbobenzyloxy-l-threonine methyl ester,z-l-threonine methyl ester,n-carbobenzoxy-l-threonine methyl ester,n-alpha-carbobenzoxy-l-threonine methyl ester,methyl 2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanoate,z-thr-ome,n-benzyloxycarbonyl threonine methyl ester,methyl 2-benzyloxy carbonyl amino-3-hydroxybutanoate #
2733409
methyl (2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoate
CC(C(C(=O)OC)NC(=O)OCC1=CC=CC=C1)O
Beilstein 2220800
Formula Weight 267.28
Physical Form Powder
Percent Purity 97%
Color White
Melting Point 92°C to 94°C
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-L-threonine methyl ester

storageNote1 : -30° to -10°C