Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-threonylglycine ethyl ester, 97%

Click to view available options
Quantity:
1g
5g
27482-74-2
C16H22N2O6
338.36
MFCD00191160
HJLLTKBIYMYCMV-RISCZKNCSA-N
z-thr-gly-oet,ethyl 2-2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanamido acetate,ethyl 2-2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanamido acetate
7019127
ethyl 2-[[(2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
CCOC(=O)CNC(=O)C(C(C)O)NC(=O)OCC1=CC=CC=C1
27482-74-2
C16H22N2O6
338.36
MFCD00191160
HJLLTKBIYMYCMV-RISCZKNCSA-N
z-thr-gly-oet,ethyl 2-2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanamido acetate,ethyl 2-2s,3r-2-benzyloxy carbonyl amino-3-hydroxybutanamido acetate
7019127
ethyl 2-[[(2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoyl]amino]acetate
CCOC(=O)CNC(=O)C(C(C)O)NC(=O)OCC1=CC=CC=C1
Beilstein 5131540
Formula Weight 338.36
Physical Form Solid
Percent Purity 97%
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-L-threonylglycine ethyl ester