Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-tryptophan benzyl ester, 95%

Click to view available options
Quantity:
1g
5g
69876-37-5
C26H24N2O4
428.488
MFCD00191163
UHYCVEDLXBEKPC-DEOSSOPVSA-N
z-trp-obzl,nalpha-cbz-l-tryptophan benzyl ester,benzyl 2s-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoate,z-l-trp-obzl,s-benzyl 2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate,benzyl 2s-2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate
14102143
benzyl (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoate
C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)OCC4=CC=CC=C4
69876-37-5
C26H24N2O4
428.488
MFCD00191163
UHYCVEDLXBEKPC-DEOSSOPVSA-N
z-trp-obzl,nalpha-cbz-l-tryptophan benzyl ester,benzyl 2s-2-benzyloxy carbonyl amino-3-1h-indol-3-yl propanoate,z-l-trp-obzl,s-benzyl 2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate,benzyl 2s-2-benzyloxycarbonylamino-3-1h-indol-3-yl propanoate
14102143
benzyl (2S)-3-(1H-indol-3-yl)-2-(phenylmethoxycarbonylamino)propanoate
C1=CC=C(C=C1)COC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)OCC4=CC=CC=C4
Beilstein 4725277
Formula Weight 428.49
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Benzyloxycarbonyl-L-tryptophan benzyl ester

storageNote1 : 2° to 8°C