Alfa Aesar™ N-Benzyloxycarbonyl-L-tyrosine, 99%, may contain up to »10% water

Manufacturer: Alfa Aesar™ A1096614

Quantity 25g

 View more versions of this product

Catalog No. AAA1096614

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1164-16-5
    Molecular Formula C17H17NO5
    Formula Weight 315.33
    MDL Number MFCD00037180
    Beilstein 2169918
    Synonym N-Cbz-L-tyrosine; Z-Tyr-OH
    Chemical Name or Material N-Benzyloxycarbonyl-L-tyrosine

    Specifications

    Formula Weight 315.33
    Melting Point 57° to 60°C
    MDL Number MFCD00037180
    CAS 1164-16-5
    Synonym N-Cbz-L-tyrosine; Z-Tyr-OH
    Optical Rotation +10° (c=3 in dioxane)
    Recommended Storage 2°C to 8°C
    Molecular Formula C17H17NO5
    Concentration May contain up to ≈10% water
    Beilstein 2169918
    Quantity 25g
    Chemical Name or Material N-Benzyloxycarbonyl-L-tyrosine
    Assay 99%

    2°C to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit