Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-L-valinamide, 97%

Click to view available options
Quantity:
5g
25g
13139-28-1
C13H18N2O3
250.298
MFCD00191170
LWAHIZLDUQONHV-NSHDSACASA-N
z-val-nh2,z-l-valine amide,s-benzyl 1-amino-3-methyl-1-oxobutan-2-yl carbamate,benzyl n-1s-1-carbamoyl-2-methylpropyl carbamate,n-cbz-l-valinamide
10562630
benzyl N-[(2S)-1-amino-3-methyl-1-oxobutan-2-yl]carbamate
CC(C)C(C(=O)N)NC(=O)OCC1=CC=CC=C1
13139-28-1
C13H18N2O3
250.298
MFCD00191170
LWAHIZLDUQONHV-NSHDSACASA-N
z-val-nh2,z-l-valine amide,s-benzyl 1-amino-3-methyl-1-oxobutan-2-yl carbamate,benzyl n-1s-1-carbamoyl-2-methylpropyl carbamate,n-cbz-l-valinamide
10562630
benzyl N-[(2S)-1-amino-3-methyl-1-oxobutan-2-yl]carbamate
CC(C)C(C(=O)N)NC(=O)OCC1=CC=CC=C1
Quantity 5g
Formula Weight 250.3
Physical Form Solid
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-L-valinamide

storageNote1 : -30° to -10°C