Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-D-alanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
68223-03-0
C12H15NO4
237.255
MFCD00153387
QGEQKVZQPWSOTI-SECBINFHSA-N
z-d-meala-oh,z-n-me-d-ala-oh,n-methyl-n-cbz-d-alanine,ambotzzaa1076,z-d-n-me-ala-oh,n-alpha-benzyloxycarbonyl-n-alpha-methyl-d-alanine,2r-2-methyl phenylmethoxycarbonyl amino propanoic acid,2r-2-benzyloxy carbonyl methyl amino propanoic acid
6994931
(2R)-2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
68223-03-0
C12H15NO4
237.255
MFCD00153387
QGEQKVZQPWSOTI-SECBINFHSA-N
z-d-meala-oh,z-n-me-d-ala-oh,n-methyl-n-cbz-d-alanine,ambotzzaa1076,z-d-n-me-ala-oh,n-alpha-benzyloxycarbonyl-n-alpha-methyl-d-alanine,2r-2-methyl phenylmethoxycarbonyl amino propanoic acid,2r-2-benzyloxy carbonyl methyl amino propanoic acid
6994931
(2R)-2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
Beilstein 6807578
Formula Weight 237.26
Physical Form Solid
Percent Purity 95%
Melting Point 215°C to 219°C
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-D-alanine

storageNote1 : 2° to 8°C