Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-D-valine, 95%

Click to view available options
Quantity:
250mg
1g
5g
53978-73-7
C14H19NO4
265.309
MFCD04973293
NNEHOKZDWLJKHP-GFCCVEGCSA-N
z-d-n-me-val-oh,n-cbz-n-methyl-d-valine,r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid,ambotzzaa1004,benzyloxycarbonyl-n-alpha-methyl-d-valine,n-benzyloxy carbonyl-n-methyl-d-valine,d-valine, n-methyl-n-phenylmethoxy carbonyl,2r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid
7018026
(2R)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
53978-73-7
C14H19NO4
265.309
MFCD04973293
NNEHOKZDWLJKHP-GFCCVEGCSA-N
z-d-n-me-val-oh,n-cbz-n-methyl-d-valine,r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid,ambotzzaa1004,benzyloxycarbonyl-n-alpha-methyl-d-valine,n-benzyloxy carbonyl-n-methyl-d-valine,d-valine, n-methyl-n-phenylmethoxy carbonyl,2r-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid
7018026
(2R)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
Beilstein 3593385
Formula Weight 265.31
Physical Form Solid
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-D-valine

storageNote1 : 2° to 8°C