Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-DL-alanine dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
250mg
1g
5g
91738-83-9
C24H38N2O4
418.578
MFCD00672350
RHGOEFBRIGKCFJ-UHFFFAOYSA-N
z-n-me-dl-ala-oh dcha,2-benzyloxy carbonyl methyl amino propanoic acid; dicha,dicyclohexylamine 2-benzyloxy carbonyl methyl amino propanoate
56777042
N-cyclohexylcyclohexanamine;2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
91738-83-9
C24H38N2O4
418.578
MFCD00672350
RHGOEFBRIGKCFJ-UHFFFAOYSA-N
z-n-me-dl-ala-oh dcha,2-benzyloxy carbonyl methyl amino propanoic acid; dicha,dicyclohexylamine 2-benzyloxy carbonyl methyl amino propanoate
56777042
N-cyclohexylcyclohexanamine;2-[methyl(phenylmethoxycarbonyl)amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1.C1CCC(CC1)NC2CCCCC2
Formula Weight 418.58
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-DL-alanine dicyclohexylammonium salt

storageNote1 : 2° to 8°C