Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-N-methyl-L-valine, 95%

Click to view available options
Quantity:
250mg
1g
5g
42417-65-2
C14H19NO4
265.309
MFCD00153397
NNEHOKZDWLJKHP-LBPRGKRZSA-N
cbz-n-methyl-l-valine,z-n-me-val-oh,cbz-n-me-val-oh,z-n-methyl-l-valine,s-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid,z-meval-oh,ambotzzaa1019,cbz-n-d-me-val-oh,cbz-n-methyl-l-val-oh,z-n-,a-methyl-l-valine
7016371
(2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
42417-65-2
C14H19NO4
265.309
MFCD00153397
NNEHOKZDWLJKHP-LBPRGKRZSA-N
cbz-n-methyl-l-valine,z-n-me-val-oh,cbz-n-me-val-oh,z-n-methyl-l-valine,s-2-benzyloxy carbonyl methyl amino-3-methylbutanoic acid,z-meval-oh,ambotzzaa1019,cbz-n-d-me-val-oh,cbz-n-methyl-l-val-oh,z-n-,a-methyl-l-valine
7016371
(2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1=CC=CC=C1
Beilstein 1990143
Formula Weight 265.31
Physical Form Crystalline Solid
Percent Purity 95%
Color White
Melting Point 68°C to 70°C
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-N-methyl-L-valine