Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-O-benzyl-D-tyrosine, 95%

Click to view available options
Quantity:
5g
25g
92455-53-3
C24H23NO5
405.45
MFCD00153349
IPAODWFPTVIUSZ-JOCHJYFZSA-N
z-d-tyr bzl-oh,cbz-o-benzyl-d-tyr,2r-2-phenylmethoxycarbonylamino-3-4-phenylmethoxyphenyl propanoic acid,ambotzzaa1214,n-alpha-benzyloxycarbonyl-o-benzyl-d-tyrosine,2r-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid
7408285
(2R)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3=CC=CC=C3
92455-53-3
C24H23NO5
405.45
MFCD00153349
IPAODWFPTVIUSZ-JOCHJYFZSA-N
z-d-tyr bzl-oh,cbz-o-benzyl-d-tyr,2r-2-phenylmethoxycarbonylamino-3-4-phenylmethoxyphenyl propanoic acid,ambotzzaa1214,n-alpha-benzyloxycarbonyl-o-benzyl-d-tyrosine,2r-2-benzyloxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid
7408285
(2R)-2-(phenylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3=CC=CC=C3
Beilstein 6941737
Formula Weight 405.45
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 113°C to 119°C
Quantity 5g
Chemical Name or Material N-Benzyloxycarbonyl-O-benzyl-D-tyrosine

storageNote1 : -30° to -10°C