Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
5g
25g
16966-07-7
C28H46N2O5
490.685
MFCD00077043
STGGZKHUOOUVBV-YLAFAASESA-N
z-thr tbu-oh.dcha,z-thr tbu-oh dcha,dicyclohexylamine 2s,3r-2-benzyloxy carbonyl amino-3-tert-butoxy butanoate,o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-l-theronine,cbz-o-tert-butyl-l-threonine dicyclohexylamine,pubchem18369,z-thr tbu-oh?cha,cbz-thr tbu-oh.dcha,z-dl-thr tbu-oh.dcha,cbz-o-tert-butyl-l-threonine dcha
15601090
N-cyclohexylcyclohexanamine;(2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)butanoic acid
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
16966-07-7
C28H46N2O5
490.685
MFCD00077043
STGGZKHUOOUVBV-YLAFAASESA-N
z-thr tbu-oh.dcha,z-thr tbu-oh dcha,dicyclohexylamine 2s,3r-2-benzyloxy carbonyl amino-3-tert-butoxy butanoate,o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-l-theronine,cbz-o-tert-butyl-l-threonine dicyclohexylamine,pubchem18369,z-thr tbu-oh?cha,cbz-thr tbu-oh.dcha,z-dl-thr tbu-oh.dcha,cbz-o-tert-butyl-l-threonine dcha
15601090
N-cyclohexylcyclohexanamine;(2S,3R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)butanoic acid
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OC(C)(C)C.C1CCC(CC1)NC2CCCCC2
Formula Weight 490.69
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-threonine dicyclohexylammonium salt

storageNote1 : -30° to -10°C