Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester, 95%

Click to view available options
Quantity:
1g
5g
25g
5068-29-1
C22H27NO5
385.46
MFCD00237370
IPRNNTJYTNRIDM-IBGZPJMESA-N
z-tyr tbu-ome,z-tyr but-ome,z-o-tert-butyl-l-tyrosine methyl ester,cbz-o-tert-butyl-l-tyrosine methyl ester,n-benzyloxycarbonyl-o-tert-butyl-l-tyrosine methyl ester,l-tyrosine,o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-, methyl ester,methyl 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate
7019804
methyl (2S)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]-2-(phenylmethoxycarbonylamino)propanoate
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC)NC(=O)OCC2=CC=CC=C2
5068-29-1
C22H27NO5
385.46
MFCD00237370
IPRNNTJYTNRIDM-IBGZPJMESA-N
z-tyr tbu-ome,z-tyr but-ome,z-o-tert-butyl-l-tyrosine methyl ester,cbz-o-tert-butyl-l-tyrosine methyl ester,n-benzyloxycarbonyl-o-tert-butyl-l-tyrosine methyl ester,l-tyrosine,o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-, methyl ester,methyl 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate
7019804
methyl (2S)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]-2-(phenylmethoxycarbonylamino)propanoate
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC)NC(=O)OCC2=CC=CC=C2
Beilstein 2676239
Formula Weight 385.46
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine methyl ester

storageNote1 : -30° to -10°C