Alfa Aesar™ N-(Benzyloxycarbonyloxy)succinimide, 99%

Manufacturer: Alfa Aesar™ A1215322

Quantity 100g

 View more versions of this product

Catalog No. AAA1215322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13139-17-8
    Molecular Formula C12H11NO5
    Formula Weight 249.23
    MDL Number MFCD00005513
    Beilstein 1387927
    Synonym Benzyl N-succinimidyl carbonate; Cbz-ONSu
    Chemical Name or Material N- (Benzyloxycarbonyloxy)succinimide

    Specifications

    Formula Weight 249.23
    CAS 13139-17-8
    Melting Point 77° to 82°C
    Quantity 100g
    Molecular Formula C12H11NO5
    MDL Number MFCD00005513
    Recommended Storage 2°C to 8°C
    Beilstein 1387927
    Synonym Benzyl N-succinimidyl carbonate; Cbz-ONSu
    Chemical Name or Material N- (Benzyloxycarbonyloxy)succinimide
    Assay 99%

    Description

    Applications
    N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

    Solubility
    Soluble in chloroform, methanol, acetone (0.1 g/mL).

    Notes
    Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.

    2°C to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit