Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylquininium chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7035700
Click to view available options
Quantity:
1 g
5 g
67174-25-8
C27H31ClN2O2
451.007
MFCD00198105
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
n-benzylquininiumchloride chiralphase-transfercatalyst
133611806
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]
67174-25-8
C27H31ClN2O2
451.007
MFCD00198105
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
n-benzylquininiumchloride chiralphase-transfercatalyst
133611806
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C to 205°C (decomposition)
Quantity 1 g
Optical Rotation −235° (c=1.5 in Water)
Formula Weight 451.01
Percent Purity 95%
Chemical Name or Material N-Benzylquininium chloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit