Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyltropinone, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
10g
28957-72-4
C14H17NO
215.30
MFCD00151640
RSUHKGOVXMXCND-UHFFFAOYNA-N
8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
119846
8-benzyl-8-azabicyclo[3.2.1]octan-3-one
O=C1CC2CCC(C1)N2CC1=CC=CC=C1
28957-72-4
C14H17NO
215.30
MFCD00151640
RSUHKGOVXMXCND-UHFFFAOYNA-N
8-benzyl-8-azabicyclo 3.2.1 octan-3-one, n-benzylnortropinone, n-benzyltropinone, 8-benzyl-8-aza-bicyclo 3.2.1 octan-3-one, 8-phenylmethyl-8-azabicyclo 3.2.1 octan-3-one, 8-azabicyclo 3.2.1 octan-3-one, 8-phenylmethyl, n-benzyl tropinone, 8-benzyl-8-azabicyclo 3.2.1 octan-3-on, n-benzyl-8-azabicyclo 3.2.1 octan-3-one
119846
8-benzyl-8-azabicyclo[3.2.1]octan-3-one
O=C1CC2CCC(C1)N2CC1=CC=CC=C1
Assay Percent Range 97%
Quantity 10g
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger