Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
286961-14-6
C16H28BNO4
309.21
VVDCRJGWILREQH-UHFFFAOYSA-N
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate,1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine,n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate,3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester,n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine,tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
4642098
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OC(C)(C)C
286961-14-6
C16H28BNO4
309.21
VVDCRJGWILREQH-UHFFFAOYSA-N
n-boc-1,2,5,6-tetrahydropyridine-4-boronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate,1-n-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine,n-boc-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate,tert-butyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate,3,6-dihydro-2h-pyridine-1-n-boc-4-boronic acid pinacol ester,n-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester,1-boc-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine,tert-butyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2,3,6-tetrahydropyridine-1-carboxylate
4642098
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OC(C)(C)C
Quantity 5g
Packaging Glass Bottle
Melting Point 115.0°C to 117.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 309.21
Percent Purity 97%
Chemical Name or Material N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid Pinacol Ester