Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-1,2-diaminoethane, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
57260-73-8
C7H16N2O2
160.22
MFCD00191871
AOCSUUGBCMTKJH-UHFFFAOYSA-N
n-boc-ethylenediamine,tert-butyl n-2-aminoethyl carbamate,tert-butyl 2-aminoethyl carbamate,n-t-boc-ethylenediamine,n-tert-boc-ethylenediamine,tert-butyl 2-aminoethylcarbamate,n-boc-1,2-diaminoethane,n-boc-ethane-1,2-diamine,mono-n-t-boc-eda,n-boc-diaminoethane
187201
tert-butyl N-(2-aminoethyl)carbamate
CC(C)(C)OC(=O)NCCN
57260-73-8
C7H16N2O2
160.22
MFCD00191871
AOCSUUGBCMTKJH-UHFFFAOYSA-N
n-boc-ethylenediamine,tert-butyl n-2-aminoethyl carbamate,tert-butyl 2-aminoethyl carbamate,n-t-boc-ethylenediamine,n-tert-boc-ethylenediamine,tert-butyl 2-aminoethylcarbamate,n-boc-1,2-diaminoethane,n-boc-ethane-1,2-diamine,mono-n-t-boc-eda,n-boc-diaminoethane
187201
tert-butyl N-(2-aminoethyl)carbamate
CC(C)(C)OC(=O)NCCN
Refractive Index 1.458
Density 1.0160g/mL
Boiling Point 80°C to 84°C (0.3mmHg)
Flash Point >110°C
Specific Gravity 1.016
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)3COC(O)NH(CH2)2NH2
Formula Weight 160.22
Physical Form Oily Liquid
Percent Purity 98%
Chemical Name or Material N-BOC-1, 2-diaminoethane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger