Alfa Aesar™ N-Boc-1,3-diaminopropane, 95%

Manufacturer: Alfa Aesar™ H5030406

Quantity 5g

 View more versions of this product

Catalog No. AAH5030406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75178-96-0
    Molecular Formula C8H18N2O2
    Linear Formula (CH3)3COCONHCH2CH2CH2NH2
    Formula Weight 174.24
    MDL Number MFCD00210021
    UN Number UN3259
    Synonym N-Boc-1,3-propanediamine; N-tert-Butoxycarbonyl-1,3-diaminopropane
    Chemical Name or Material N-Boc-1,3-diaminopropane

    Specifications

    Formula Weight 174.24
    Molecular Formula C8H18N2O2
    Linear Formula (CH3)3COCONHCH2CH2CH2NH2
    Melting Point ≈22°C
    MDL Number MFCD00210021
    CAS 75178-96-0
    UN Number UN3259
    Quantity 5g
    Synonym N-Boc-1,3-propanediamine; N-tert-Butoxycarbonyl-1,3-diaminopropane
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
    Chemical Name or Material N-Boc-1,3-diaminopropane
    Assay 95%

    Description

    Applications
    N-Boc-1,3-diaminopropane is used in the preparation of spermidine analogues as well as in the preparation of pharmacologically active compounds. It is also used in suzuki reaction. Further, it is used in the preparation of [3-(3-cyano-propylamino)-propyl]-carbamic acid tert-butyl ester.

    Solubility
    Miscible with methanol.

    Notes
    Hygroscopic. Air sensitive. Incompatible with oxidizing agents and acids.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit