Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-1,3-diaminopropane, 97%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
75178-96-0
C8H18N2O2
174.24
POHWAQLZBIMPRN-UHFFFAOYSA-N
n-boc-1,3-propanediamine,n-boc-1,3-diaminopropane,tert-butyl n-3-aminopropyl carbamate,tert-butyl 3-aminopropyl carbamate,n-3-aminopropyl carbamic acid tert-butyl ester,tert-butyl 3-aminopropylcarbamate,1-boc-amino-1,3-propanediamine,3-amino-propyl-carbamic acid tert-butyl ester,n-t-boc-1,3-propanediamine,n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCCN
75178-96-0
C8H18N2O2
174.24
POHWAQLZBIMPRN-UHFFFAOYSA-N
n-boc-1,3-propanediamine,n-boc-1,3-diaminopropane,tert-butyl n-3-aminopropyl carbamate,tert-butyl 3-aminopropyl carbamate,n-3-aminopropyl carbamic acid tert-butyl ester,tert-butyl 3-aminopropylcarbamate,1-boc-amino-1,3-propanediamine,3-amino-propyl-carbamic acid tert-butyl ester,n-t-boc-1,3-propanediamine,n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCCN
Specific Gravity 0.998
Boiling Point 94.0°C to 96.0°C (0.2 Torr)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 22.0°C
Density 0.9980g/mL
Quantity 5mL
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2
Formula Weight 174.24
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material N-BOC-1, 3-diaminopropane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger