Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-1,3-propanediamine hydrochloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
127346-48-9
C8H18N2O2·HCl
210.71
MFCD01076598
WUXOJNUZYOFBMI-UHFFFAOYSA-N
tert-butyl 3-aminopropyl carbamate hydrochloride,n-boc-1,3-diaminopropane hydrochloride,n-3-aminopropyl carbamic acid tert-butyl ester hydrochloride,n-boc-1,3-propanediamine hydrochloride,tert-butyl n-3-aminopropyl carbamate hydrochloride,acmc-20ak6b,n-boc-1,3-diaminopropane hcl,n-3-aminopropyl carbamic acid tert-butyl ester-hcl,tert-butyl 3-aminopropyl carbamatehydrochloride,n-3-aminopropyl carbamic acid tert-butyl ester hcl
21982912
tert-butyl N-(3-aminopropyl)carbamate;hydrochloride
CC(C)(C)OC(=O)NCCCN.Cl
127346-48-9
C8H18N2O2·HCl
210.71
MFCD01076598
WUXOJNUZYOFBMI-UHFFFAOYSA-N
tert-butyl 3-aminopropyl carbamate hydrochloride,n-boc-1,3-diaminopropane hydrochloride,n-3-aminopropyl carbamic acid tert-butyl ester hydrochloride,n-boc-1,3-propanediamine hydrochloride,tert-butyl n-3-aminopropyl carbamate hydrochloride,acmc-20ak6b,n-boc-1,3-diaminopropane hcl,n-3-aminopropyl carbamic acid tert-butyl ester-hcl,tert-butyl 3-aminopropyl carbamatehydrochloride,n-3-aminopropyl carbamic acid tert-butyl ester hcl
21982912
tert-butyl N-(3-aminopropyl)carbamate;hydrochloride
CC(C)(C)OC(=O)NCCCN.Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2·HCl
Formula Weight 210.71
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material N-BOC-1, 3-propanediamine hydrochloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning