Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-1,4-diaminocyclohexane, 95%

Click to view available options
Quantity:
250mg
1g
5g
195314-59-1
C11H22N2O2
214.309
MFCD01076211
FEYLUKDSKVSMSZ-UHFFFAOYSA-N
trans-n-boc-1,4-cyclohexanediamine,n-boc-trans-1,4-cyclohexanediamine,1-n-boc-cis-1,4-cyclohexyldiamine,tert-butyl 4-aminocyclohexyl carbamate,n-boc-1,4-cyclohexanediamine,tert-butyl n-4-aminocyclohexyl carbamate,tert-butyl cis-4-aminocyclohexyl carbamate,1-boc-amino-1,4-cyclohexanediamine,tert-butyl 4-aminocyclohexylcarbamate,cis-n-boc-1,4-cyclohexanediamine
2756050
tert-butyl N-(4-aminocyclohexyl)carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)N
195314-59-1
C11H22N2O2
214.309
MFCD01076211
FEYLUKDSKVSMSZ-UHFFFAOYSA-N
trans-n-boc-1,4-cyclohexanediamine,n-boc-trans-1,4-cyclohexanediamine,1-n-boc-cis-1,4-cyclohexyldiamine,tert-butyl 4-aminocyclohexyl carbamate,n-boc-1,4-cyclohexanediamine,tert-butyl n-4-aminocyclohexyl carbamate,tert-butyl cis-4-aminocyclohexyl carbamate,1-boc-amino-1,4-cyclohexanediamine,tert-butyl 4-aminocyclohexylcarbamate,cis-n-boc-1,4-cyclohexanediamine
2756050
tert-butyl N-(4-aminocyclohexyl)carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)N
Quantity 250mg
Color White
UN Number UN3259
Beilstein 13428745
Formula Weight 214.31
Physical Form Solid
Percent Purity 95%
Chemical Name or Material N-Boc-1,4-diaminocyclohexane

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.