Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-BOC-1-Aminocyclobutanecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
120728-10-1
C10H17NO4
215.25
ROVVUKFHORPDSM-UHFFFAOYSA-N
n-boc-1-aminocyclobutanecarboxylic acid,1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid,1-boc-amino cyclobutanecarboxylic acid,boc-cyclovaline,boc-acbc-oh,1-n-boc-amino-cyclobutane carboxylic acid,1-tert-butoxycarbonylamino cyclobutanecarboxylic acid,boc-1-aminocyclobutane-1-carboxylic acid,n-boc-1-aminocyclobutane carboxylic acid,boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
120728-10-1
C10H17NO4
215.25
ROVVUKFHORPDSM-UHFFFAOYSA-N
n-boc-1-aminocyclobutanecarboxylic acid,1-tert-butoxycarbonyl amino cyclobutanecarboxylic acid,1-boc-amino cyclobutanecarboxylic acid,boc-cyclovaline,boc-acbc-oh,1-n-boc-amino-cyclobutane carboxylic acid,1-tert-butoxycarbonylamino cyclobutanecarboxylic acid,boc-1-aminocyclobutane-1-carboxylic acid,n-boc-1-aminocyclobutane carboxylic acid,boc-1-aminocyclobutanecarboxylic acid
1512646
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCC1)C(=O)O
Melting Point 129°C to 133°C
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Color White to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 215.25
Physical Form Powder
Percent Purity 97%
Chemical Name or Material N-BOC-1-Aminocyclobutanecarboxylic acid
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you fee

GHS Signal Word: Warning