Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-2-(2-naphthyl)-D-alanine, 99%

Manufacturer:  Alfa Aesar™ H5196803

Quantity 1g

 View more versions of this product

Catalog No. AAH5196803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

76985-10-9
315.37
4200660
N-Boc-2-(2-naphthyl)-D-alanine
C18H21NO4
MFCD00076900
Boc-D-2-Nal-OH

Specifications

76985-10-9
315.37
MFCD00076900
1g
−45° (c=1 in Ethanol)
99%
C18H21NO4
94° to 96°C
4200660
Boc-D-2-Nal-OH
N-Boc-2-(2-naphthyl)-D-alanine
missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'