Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-2-(3-pyridyl)-D-alanine, 95%

Manufacturer:  Alfa Aesar™ H5198503

Quantity 1g

 View more versions of this product

Catalog No. AAH5198503


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

98266-33-2
266.3
4195466
N-Boc-2-(3-pyridyl)-D-alanine
C13H18N2O4
MFCD00076901
Boc-D-3-Pal-OH

Specifications

98266-33-2
266.3
4195466
Boc-D-3-Pal-OH
95%
C13H18N2O4
MFCD00076901
1g
N-Boc-2-(3-pyridyl)-D-alanine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit