Alfa Aesar™ N-Boc-2-(3-pyridyl)-D-alanine, 95%

Manufacturer: Alfa Aesar™ H51985MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51985MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98266-33-2
    Molecular Formula C13H18N2O4
    Formula Weight 266.3
    MDL Number MFCD00076901
    Beilstein 4195466
    Synonym Boc-D-3-Pal-OH
    Chemical Name or Material N-Boc-2-(3-pyridyl)-D-alanine

    Specifications

    Formula Weight 266.3
    MDL Number MFCD00076901
    Quantity 250mg
    Molecular Formula C13H18N2O4
    CAS 98266-33-2
    Beilstein 4195466
    Synonym Boc-D-3-Pal-OH
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material N-Boc-2-(3-pyridyl)-D-alanine
    Assay 95%

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit