Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
250mg
1g
5g
676559-50-5
C12H21NO4
243.303
MFCD05663437
WBSJQVRMQOLSAT-VIFPVBQESA-N
boc-l-cyclopentylglycine,s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid,s-tert-butoxycarbonylamino-cyclopentyl-acetic acid,s-tert-butoxycarbonyl amino cyclopentyl acetic acid,2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid,2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid,ambotzbaa5140,boc-l-cyclopentyl glycine,n-tert-butoxycarbonyl-l-cyclopentylglycine,n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine
7020746
(2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1CCCC1)C(=O)O
676559-50-5
C12H21NO4
243.303
MFCD05663437
WBSJQVRMQOLSAT-VIFPVBQESA-N
boc-l-cyclopentylglycine,s-2-tert-butoxycarbonyl amino-2-cyclopentylacetic acid,s-tert-butoxycarbonylamino-cyclopentyl-acetic acid,s-tert-butoxycarbonyl amino cyclopentyl acetic acid,2s-tert-butoxycarbonyl amino cyclopentyl ethanoic acid,2s-2-tert-butoxy carbonyl amino-2-cyclopentylacetic acid,ambotzbaa5140,boc-l-cyclopentyl glycine,n-tert-butoxycarbonyl-l-cyclopentylglycine,n-alpha-t-butyloxycarbonyl-l-cyclopentylglycine
7020746
(2S)-2-cyclopentyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1CCCC1)C(=O)O
Formula Weight 424.62
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material N-Boc-2-cyclopentyl-L-glycine dicyclohexylammonium salt

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C