Alfa Aesar™ N-Boc-2-methyl-L-proline, 97%

Manufacturer: Alfa Aesar™ H52796MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52796MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 103336-06-7
    Molecular Formula C11H19NO4
    Formula Weight 229.28
    MDL Number MFCD08706361
    Chemical Name or Material N-Boc-2-methyl-L-proline

    Specifications

    Formula Weight 229.28
    Molecular Formula C11H19NO4
    MDL Number MFCD08706361
    CAS 103336-06-7
    Quantity 250mg
    Chemical Name or Material N-Boc-2-methyl-L-proline
    Assay 97%

    Description

    Applications
    N-Boc-2-methyl-L-proline is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit