Alfa Aesar™ N-Boc-2-propargyl-L-glycine, 95%

Manufacturer: Alfa Aesar™ H5212803

Quantity 1g

 View more versions of this product

Catalog No. AAH5212803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63039-48-5
    Molecular Formula C10H15NO4
    Formula Weight 213.23
    MDL Number MFCD01320855
    Chemical Name or Material N-Boc-2-propargyl-L-glycine

    Specifications

    Formula Weight 213.23
    Molecular Formula C10H15NO4
    MDL Number MFCD01320855
    CAS 63039-48-5
    Quantity 1g
    Chemical Name or Material N-Boc-2-propargyl-L-glycine
    Assay 95%

    Description

    Applications
    N-Boc-2-propargyl-L-glycine is used as pharmaceutical intermediate.

    Solubility
    Soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit