Alfa Aesar™ N-Boc-3-(2-thienyl)-L-alanine, 95%

Manufacturer: Alfa Aesar™ H51971MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51971MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56675-37-7
    Molecular Formula C12H17NO4S
    Formula Weight 271.33
    MDL Number MFCD00062051
    Beilstein 1388299
    Synonym Boc-beta-(2-thienyl)-Ala-OH
    Chemical Name or Material N-Boc-3-(2-thienyl)-L-alanine

    Specifications

    Formula Weight 271.33
    Melting Point 71° to 76°C
    MDL Number MFCD00062051
    Quantity 250mg
    Optical Rotation +11.0° (c=1 in Methanol)
    Molecular Formula C12H17NO4S
    CAS 56675-37-7
    Beilstein 1388299
    Synonym Boc-beta-(2-thienyl)-Ala-OH
    Chemical Name or Material N-Boc-3-(2-thienyl)-L-alanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit