Alfa Aesar™ N-Boc-3,4-dichloro-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H5205306

Quantity 5g

 View more versions of this product

Catalog No. AAH5205306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80741-39-5
    Molecular Formula C14H17Cl2NO4
    Formula Weight 334.2
    MDL Number MFCD00273436
    Beilstein 6426101
    Synonym Boc-Phe(3,4-Cl2)-OH
    Chemical Name or Material N-Boc-3,4-dichloro-L-phenylalanine

    Specifications

    Formula Weight 334.2
    Molecular Formula C14H17Cl2NO4
    MDL Number MFCD00273436
    CAS 80741-39-5
    Beilstein 6426101
    Quantity 5g
    Synonym Boc-Phe(3,4-Cl2)-OH
    Chemical Name or Material N-Boc-3,4-dichloro-L-phenylalanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit