Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-3,4-difluoro-D-phenylalanine, 98%

Click to view available options
Quantity:
1g
5g
205445-51-8
C14H17F2NO4
301.29
MFCD00672519
CYAOPVHXASZUDE-LLVKDONJSA-N
boc-d-3,4-difluorophenylalanine,boc-3,4-difluoro-d-phenylalanine,boc-d-phe 3,4-dif-oh,boc-d-3,4-difluorophe,boc-d-phe 3,4-f2-oh,n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine,2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid,n-boc-3,4-difluoro-d-phenylalanine,n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine,3,4-difluoro-d-phenylalanine, boc protected
2736194
(2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O
205445-51-8
C14H17F2NO4
301.29
MFCD00672519
CYAOPVHXASZUDE-LLVKDONJSA-N
boc-d-3,4-difluorophenylalanine,boc-3,4-difluoro-d-phenylalanine,boc-d-phe 3,4-dif-oh,boc-d-3,4-difluorophe,boc-d-phe 3,4-f2-oh,n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine,2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid,n-boc-3,4-difluoro-d-phenylalanine,n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine,3,4-difluoro-d-phenylalanine, boc protected
2736194
(2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O
Beilstein 1920842
Formula Weight 301.29
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material N-Boc-3,4-difluoro-D-phenylalanine

storageNote1 : 2° to 8°C