Alfa Aesar™ N-Boc-3-bromo-2-isoxazoline-5-methylamine, 97%

Manufacturer: Alfa Aesar™ H2627427

Quantity 2.5g

 View more versions of this product

Catalog No. AAH2627427

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109770-82-3
    Molecular Formula C9H15BrN2O3
    Formula Weight 279.14
    MDL Number MFCD08741480
    Synonym (3-Bromo-4,5-dihydro-5-isoxazolylmethyl)carbamic acid tert-butyl ester; N-tert-Butoxycarbonyl-3-bromo-2-isoxazolidine-5-methylamine
    Chemical Name or Material N-Boc-3-bromo-2-isoxazoline-5-methylamine

    Specifications

    Formula Weight 279.14
    Quantity 2.5g
    Synonym (3-Bromo-4,5-dihydro-5-isoxazolylmethyl)carbamic acid tert-butyl ester; N-tert-Butoxycarbonyl-3-bromo-2-isoxazolidine-5-methylamine
    Molecular Formula C9H15BrN2O3
    MDL Number MFCD08741480
    CAS 109770-82-3
    Chemical Name or Material N-Boc-3-bromo-2-isoxazoline-5-methylamine
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit