Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-3-bromo-D-phenylalanine, 98%

Click to view available options
Quantity:
250mg
1g
5g
261360-77-4
C14H18BrNO4
344.205
MFCD01317704
FBUDYESOPLBQIR-LLVKDONJSA-N
boc-3-bromo-d-phenylalanine,r-n-boc-3-bromophenylalanine,boc-d-3-bromophenylalanine,r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-3-br-phe-oh,boc-d-3-bromophe,2r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-phe 3-br-oh,r-boc-2-amino-3-3-bromophenyl propionic acid,d-phenylalanine, 3-bromo-n-1,1-dimethylethoxy carbonyl
7006629
(2R)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O
261360-77-4
C14H18BrNO4
344.205
MFCD01317704
FBUDYESOPLBQIR-LLVKDONJSA-N
boc-3-bromo-d-phenylalanine,r-n-boc-3-bromophenylalanine,boc-d-3-bromophenylalanine,r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-3-br-phe-oh,boc-d-3-bromophe,2r-3-3-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid,boc-d-phe 3-br-oh,r-boc-2-amino-3-3-bromophenyl propionic acid,d-phenylalanine, 3-bromo-n-1,1-dimethylethoxy carbonyl
7006629
(2R)-3-(3-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)Br)C(=O)O
Formula Weight 344.2
Physical Form Powder
Percent Purity 98%
Color White
Quantity 1g
Chemical Name or Material N-Boc-3-bromo-D-phenylalanine

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold