Alfa Aesar™ N-Boc-3-bromo-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H52024MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52024MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 82278-73-7
    Molecular Formula C14H18BrNO4
    Formula Weight 344.2
    MDL Number MFCD01317710
    Synonym Boc-Phe(3-Br)-OH
    Chemical Name or Material N-Boc-3-bromo-L-phenylalanine

    Specifications

    Formula Weight 344.2
    MDL Number MFCD01317710
    Quantity 250mg
    Molecular Formula C14H18BrNO4
    CAS 82278-73-7
    Synonym Boc-Phe(3-Br)-OH
    Chemical Name or Material N-Boc-3-bromo-L-phenylalanine
    Assay 95%

    Description

    Applications
    N-Boc-3-bromo-L-phenylalanine is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit