Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-3-dimethylamino-DL-alanine, 97%

Manufacturer:  Alfa Aesar™ H5241206

Quantity 5g

 View more versions of this product

Catalog No. AAH5241206


  Check Availability
Add to cart

Specifications

Description

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

851653-36-6
232.28
(+/-)-2-(Boc-amino)-3-(dimethylamino)propionic acid; N-tert-Butoxycarbonyl-3-dimethylamino-DL-alanine
C10H20N2O4
MFCD04972270
N-Boc-3-dimethylamino-DL-alanine

Specifications

851653-36-6
232.28
5g
N-Boc-3-dimethylamino-DL-alanine
C10H20N2O4
MFCD04972270
(+/-)-2-(Boc-amino)-3-(dimethylamino)propionic acid; N-tert-Butoxycarbonyl-3-dimethylamino-DL-alanine
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit