Alfa Aesar™ N-Boc-3-fluoro-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H51988MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51988MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114873-01-7
    Molecular Formula C14H18FNO4
    Formula Weight 283.3
    MDL Number MFCD00672522
    Beilstein 5347375
    Synonym Boc-Phe(3-F)-OH
    Chemical Name or Material N-Boc-3-fluoro-L-phenylalanine

    Specifications

    Formula Weight 283.3
    Melting Point 75° to 80°C
    Quantity 250mg
    Optical Rotation +6.5° (c=1 in Methanol)
    Molecular Formula C14H18FNO4
    MDL Number MFCD00672522
    CAS 114873-01-7
    Beilstein 5347375
    Synonym Boc-Phe(3-F)-OH
    Chemical Name or Material N-Boc-3-fluoro-L-phenylalanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit