Alfa Aesar™ N-Boc-4,4-difluoro-L-proline methyl ester, 98%

Manufacturer: Alfa Aesar™ H2772703

Quantity 1g

 View more versions of this product

Catalog No. AAH2772703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 203866-17-5
    Molecular Formula C11H17F2NO4
    Formula Weight 265.26
    MDL Number MFCD03094918
    Chemical Name or Material N-Boc-4,4-difluoro-L-proline methyl ester

    Specifications

    Formula Weight 265.26
    Molecular Formula C11H17F2NO4
    MDL Number MFCD03094918
    Quantity 1g
    Recommended Storage 2° to 8°C
    CAS 203866-17-5
    Chemical Name or Material N-Boc-4,4-difluoro-L-proline methyl ester
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit