Alfa Aesar™ N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt, 97%

Manufacturer: Alfa Aesar™ H27005MF

Quantity 50mg

 View more versions of this product

Catalog No. AAH27005MF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87720-54-5
    Molecular Formula C11H19NO4·C12H23N
    Formula Weight 410.59
    MDL Number MFCD00237524
    UN Number UN3077
    Synonym (S)-2-(Boc-amino)-4-methyl-4-pentenoic acid dicyclohexylamine salt; Boc-4,5-dehydro-Leu-OH.DCHA
    Chemical Name or Material N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt

    Specifications

    Formula Weight 410.59
    Molecular Formula C11H19NO4·C12H23N
    Melting Point 148° to 153°C
    MDL Number MFCD00237524
    UN Number UN3077
    Quantity 50mg
    Synonym (S)-2-(Boc-amino)-4-methyl-4-pentenoic acid dicyclohexylamine salt; Boc-4,5-dehydro-Leu-OH.DCHA
    Optical Rotation +13° (c=1 in Methanol)
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2° to 8°C
    CAS 87720-54-5
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material N-Boc-4,5-dehydro-L-leucine dicyclohexylamine salt
    Assay 97%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit