Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-4-iodo-D-phenylalanine, 95%

Click to view available options
Quantity:
5g
25g
176199-35-2
C14H18INO4
391.205
MFCD00079670
JZLZDBGQWRBTHN-LLVKDONJSA-N
boc-d-phe 4-i-oh,boc-4-iodo-d-phenylalanine,boc-d-4-iodophenylalanine,r-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid,2r-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid,boc-4'-iodo-d-phe,n-boc-4-iodo-l-phenylalanine,ambotzbaa1280,boc-4-iodo-d-phe,boc-p-iodo-d-phe-oh
7020906
(2R)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)I)C(=O)O
176199-35-2
C14H18INO4
391.205
MFCD00079670
JZLZDBGQWRBTHN-LLVKDONJSA-N
boc-d-phe 4-i-oh,boc-4-iodo-d-phenylalanine,boc-d-4-iodophenylalanine,r-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid,2r-2-tert-butoxycarbonyl amino-3-4-iodophenyl propanoic acid,boc-4'-iodo-d-phe,n-boc-4-iodo-l-phenylalanine,ambotzbaa1280,boc-4-iodo-d-phe,boc-p-iodo-d-phe-oh
7020906
(2R)-3-(4-iodophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)I)C(=O)O
Beilstein 7518620
Formula Weight 391.21
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Sensitivity Light Sensitive
Chemical Name or Material N-Boc-4-iodo-D-phenylalanine