Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Boc-4-methyl-L-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H5198303

Quantity 1g

 View more versions of this product

Catalog No. AAH5198303


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

80102-26-7
279.34
5381557
N-Boc-4-methyl-L-phenylalanine
C15H21NO4
MFCD01317027
Boc-Phe(4-Me)-OH

Specifications

80102-26-7
279.34
MFCD01317027
1g
+16° (c=1 in Methanol)
95%
C15H21NO4
84° to 88°C
5381557
Boc-Phe(4-Me)-OH
N-Boc-4-methyl-L-phenylalanine
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit