Alfa Aesar™ N-Boc-4-methyl-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H51983MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51983MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80102-26-7
    Molecular Formula C15H21NO4
    Formula Weight 279.34
    MDL Number MFCD01317027
    Beilstein 5381557
    Synonym Boc-Phe(4-Me)-OH
    Chemical Name or Material N-Boc-4-methyl-L-phenylalanine

    Specifications

    Formula Weight 279.34
    CAS 80102-26-7
    Synonym Boc-Phe(4-Me)-OH
    Optical Rotation +16° (c=1 in Methanol)
    Molecular Formula C15H21NO4
    Melting Point 84° to 88°C
    MDL Number MFCD01317027
    Beilstein 5381557
    Quantity 250mg
    Chemical Name or Material N-Boc-4-methyl-L-phenylalanine
    Assay 95%

    Description

    Applications
    It is used as an active pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit