Alfa Aesar™ N-Boc-4-methylene-L-proline, 97%

Manufacturer: Alfa Aesar™ H2774603

Quantity 1g

 View more versions of this product

Catalog No. AAH2774603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 84348-38-9
    Molecular Formula C11H17NO4
    Formula Weight 227.26
    MDL Number MFCD01861787
    Synonym (S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
    Chemical Name or Material N-Boc-4-methylene-L-proline

    Specifications

    Formula Weight 227.26
    Molecular Formula C11H17NO4
    Melting Point 110° to 114°C
    MDL Number MFCD01861787
    Quantity 1g
    Synonym (S)-4-Methylenepyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester
    Recommended Storage 2° to 8°C
    CAS 84348-38-9
    Chemical Name or Material N-Boc-4-methylene-L-proline
    Assay 97%

    Description

    Applications
    As important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry. The fluorous prolins were synthesized from commercially available N-Boc-4-methylene- L-proline.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit